Learn More
Medchemexpress LLC 5-Bromo-1H-indole-2-carboxylic acid | 7254-19-5 | 240.06 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0075931G
Encompass_Preferred
5-Bromo-1H-indole-2-carboxylic acid is an indole-derived organic compound characterized by the presence of bromine and carboxyl groups. This compound functions as a crucial intermediate in the synthesis of various other active compounds. Additionally, its dimeric form has demonstrated antibacterial properties against Gram-positive bacteria, including Listeria monocytogenes.
  • Indole-derived organic compound
  • Substituted with bromine and carboxyl groups
  • Functions as an important intermediate for synthesis
  • Dimeric form exhibits antibacterial activity against Gram-positive bacteria
Catalog No. 50-004-48736