Learn More
Medchemexpress LLC 5-Bromo-2,3-dihydro-1H-inden-1-one | 34598-49-7 | 99.9% | 211.06 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY40190
Encompass_Preferred
5-Bromo-2,3-dihydro-1H-inden-1-one is a bromoindanone compound. It acts as a key intermediate in organic synthesis, participating in reactions such as Suzuki coupling and Buchwald amination. This compound can be utilized in the research of obesity-related metabolic diseases.
  • Key intermediate in organic synthesis
  • Participates in Suzuki coupling and Buchwald amination reactions
  • Useful in obesity-related metabolic disease research
Catalog No. 50-004-02421