Learn More
Medchemexpress LLC 5-bromo-2-chloro-4-methylpyridine | 778611-64-6 | MFCD03427663 | 98.6% | 206.47 g/mol | C6H5BrClN | 10 G

Supplier: Medchemexpress LLC HYW01458410G
5-Bromo-2-chloro-4-methylpyridine is an organic biochemical reagent supplied for research use as an intermediate and building block in medicinal chemistry and life-science applications. It is characterized with standard chemical data and a reported purity for laboratory synthesis and study.
- High reported purity of 98.62%.
- Chemical formula C6H5BrClN.
- Molecular weight 206.47 g/mol.
- Available in small laboratory pack sizes (10 g, 25 g; larger sizes by inquiry).
- Suitable as a synthetic intermediate for organic and medicinal chemistry research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.