Learn More
Medchemexpress LLC 5-Bromo-2-chloropyrimidine | 32779-36-5 | 99.9% | C4H2BrClN2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY76897
Encompass_Preferred
5-Bromo-2-chloropyrimidine is a biochemical reagent used as a biological material or organic compound for life science research. It serves as a pharmaceutical intermediate in the synthesis of pharmaceutical goods, such as inhibitors, and is also employed in cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.
  • Biochemical reagent for life science research
  • Used as a pharmaceutical intermediate
  • Employed in cross-coupling reactions
Catalog No. 50-003-58579