Learn More
Medchemexpress LLC 5-Bromo-2-chloropyrimidine | 32779-36-5 | MFCD00483232 | 99.9% | C4H2BrClN2 | 50 G
SDP
Supplier:  Medchemexpress LLC HY7689750G
Encompass_Preferred
5-Bromo-2-chloropyrimidine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used as a pharmaceutical intermediate
  • Employed in cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines
Catalog No. 50-004-33766