Learn More
Medchemexpress LLC 5-bromo-2-iodopyridine | 223463-13-6 | 283.89 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00266810G
Encompass_Preferred
5-Bromo-2-iodopyridine is a pyridine derivative containing bromine and iodine, which can be utilized in the synthesis of other active compounds, such as Lonafarnib. It can react with active zinc under mild conditions to form 5-bromo-2-pyridylzinc iodide, which is useful in various coupling reactions to obtain the corresponding cross-coupling products.
  • Pyridine derivative containing bromine and iodine
  • Can be used in the synthesis of other active compounds, such as Lonafarnib
  • Reacts with active zinc under mild conditions to form 5-bromo-2-pyridylzinc iodide
  • Useful in various coupling reactions to obtain corresponding cross-coupling products
Catalog No. 50-004-45841