Learn More
Medchemexpress LLC 5-Bromo-2-iodopyridine | 223463-13-6 | 283.89 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002668
Encompass_Preferred
5-Bromo-2-iodopyridine is a pyridine derivative that contains both bromine and iodine. This compound is utilized in the synthesis of other active compounds, such as Lonafarnib. This product is intended for research use only and is not sold to patients.
  • It can react with active zinc under mild conditions to create 5-bromo-2-pyridylzinc iodide.
  • The resulting 5-bromo-2-pyridylzinc iodide can be employed in various coupling reactions to yield corresponding cross-coupling products.
Catalog No. 50-004-48056