Learn More
Medchemexpress LLC 5-bromo-2-iodopyridine | 223463-13-6 | 283.89 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0026685G
Encompass_Preferred
5-Bromo-2-iodopyridine is a pyridine derivative containing bromine and iodine. It is used in the synthesis of other active compounds, such as Lonafarnib, and can react with active zinc under mild conditions to form 5-bromo-2-pyridylzinc iodide, which is useful in various coupling reactions.
  • Pyridine derivative containing bromine and iodine
  • Used in the synthesis of active compounds, including Lonafarnib
  • Reacts with active zinc under mild conditions to form 5-bromo-2-pyridylzinc iodide
  • Useful in various cross-coupling reactions
Catalog No. 50-004-48601