Learn More
Medchemexpress LLC 5-Bromo-2-iodopyridine | 223463-13-6 | 283.89 | 50 G
Supplier: Medchemexpress LLC HYW00266850G
5-Bromo-2-iodopyridine is a pyridine derivative containing bromine and iodine. It can be used in the synthesis of other active compounds, such as Lonafarnib. It can also react with active zinc under mild conditions to form 5-bromo-2-pyridylzinc iodide, which is utilized in various coupling reactions to obtain corresponding cross-coupling products.
- Pyridine derivative containing bromine and iodine
- Used in the synthesis of active compounds like Lonafarnib
- Reacts with active zinc to form 5-bromo-2-pyridylzinc iodide
- Utilized in various coupling reactions