Learn More
Medchemexpress LLC 5-Bromo-3-indolyl β-D-galactopyranoside | 97753-82-7 | 99.7% | 374.18 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137276100MG
Encompass_Preferred
5-Bromo-3-indolyl β-D-galactopyranoside (Bluo-Gal) is a chromogenic substrate for β-galactosidase. Upon enzymatic hydrolysis, it forms an insoluble blue precipitate, enabling specific identification of bacterial β-galactosidases (e.g., *E. coli*'s lacZ gene) at pH 7.4. It is useful for light and electron microscopy, and histochemical detection of reporter gene expression in transgenic organisms for β-galactosidase activity localization.
  • Chromogenic substrate for β-galactosidase
  • Produces an insoluble blue precipitate upon hydrolysis
  • Enables specific recognition of bacterial β-galactosidases
  • Effective at pH 7.4
  • Suitable for light and electron microscopic observations
  • Useful for histochemical detection of reporter gene expression
  • Applicable for localization analysis of β-galactosidase activity in transgenic organisms
Catalog No. 50-003-48567