Learn More
Medchemexpress LLC 5-bromo-3-indolyl β-D-galactopyranoside | 97753-82-7 | MFCD00063691 | 99.7% | 374.18 | C14H16BrNO6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1372765MG
Encompass_Preferred
5-Bromo-3-indolyl β-D-galactopyranoside is a chromogenic β-galactosidase substrate that is hydrolyzed to a 5-bromoindole intermediate which oxidizes to an insoluble blue precipitate. It is intended for histochemical detection and localization of β-galactosidase activity in biological samples and is supplied as a high-purity solid (5 mg).
  • Chromogenic substrate for β-galactosidase.
  • Generates an insoluble blue precipitate for clear localization.
  • Suitable for light and electron microscopy applications.
  • High purity for reproducible staining results.
  • Supplied as a solid for convenient handling and storage.
Catalog No. 50-002-21972