Learn More
Bioworld 5-Bromo-4-chloro-3-indolyl-phospate-P-toluidine salt (BCIP, P-Toluidine salt), 100 mg
Supplier:  Bioworld 215300341
Encompass_Preferred
Histochemical substrate for alkaline phosphataseSpecificationBCIP P-Toluidine SaltUltra Pure Grade Purity: >99%Solubility: Soluble in DMF (20mg/mL)
Catalog No. 50-198-828