Learn More
Alkali Scientific 5 Bromo 4 chloro 3 indolyl β D cellobioside [X β D Cellobioside], 100 Mg
SDP
Supplier:  Alkali Scientific OMO001CP
Encompass_Preferred
Indolyl β-D Cellobioside (5-Bromo-4-chloro-3-indolyl β-D cellobioside) is a chromogenic substrate commonly used in enzyme detection assays, especially for β-glucosidases and cellulases. This reagent contains an indole derivative that, when cleaved by appropriate enzymes, produces a colored product that can be visually detected or quantified. The 100mg quantity is ideal for small-scale assays, especially in microbiological research, where enzyme activity in bacterial or fungal cultures is assessed. Indolyl β-D cellobioside is frequently employed in the study of carbohydrate-active enzymes, as well as in screening for lignocellulose-degrading microorganisms.
Catalog No. 50-006-60002