Learn More
Bioworld 5-Bromo-4-chloro-3-indoyl-B-D-Galactoside [X-Gal] solution, 5 mL
Supplier:  Bioworld 215300362
Encompass_Preferred
Solution contains 50 mg/ml X-GAL in DMF. Inquire regarding additional concentrations. This product is sold for laboratory research use only.
Catalog No. 50-198-834