Learn More
Medchemexpress LLC 5-Bromo-L-tryptophan | 25197-99-3 | 99.8% | 283.13 | 5 G

Supplier: Medchemexpress LLC HY20897A5G
5-Bromo-L-tryptophan is an alpha-amino acid derivative found in Semenospongia sp. It is used to synthesize 5-bromoindole. This product has a molecular weight of 283.13, a formula of C11H11BrN2O2, and a CAS number of 25197-99-3. It appears as an off-white to light brown solid.
- Found in Semenospongia sp.
- Used to synthesize 5-bromoindole
- Appearance: Solid, off-white to light brown
- Initial source: Endogenous metabolite
- Shipping: Room temperature in continental US
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.