Learn More
Medchemexpress LLC 5-Bromopentanoyl chloride | 4509-90-4 | 98.63% | 199.47 | 5 G
SDP
Supplier:  Medchemexpress LLC HY435385G
Encompass_Preferred
5-Bromopentanoyl chloride is a PROTAC linker, suitable for the synthesis of PROTACs. It is found in PROTAC ATR degrader-2 (HY-161615).
  • Used as a PROTAC linker
  • Suitable for the synthesis of PROTACs
Catalog No. 50-004-03608