Learn More
Medchemexpress LLC 5-Bromotetralone | 68449-30-9 | 225.08 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0047885G
Encompass_Preferred
5-Bromotetralone (5-Bromo-3,4-dihydro-2H-naphthalen-1-one) is a synthetic intermediate useful for pharmaceutical synthesis. Storage conditions:
  • Store at room temperature for 3 years.
  • In solvent, store at -80°C for 2 years or -20°C for 1 year.
Catalog No. 50-004-48994