Learn More
Medchemexpress LLC 5-Bromothiophene-2-carbaldehyde | 4701-17-1 | 99.4% | C5H3BrOS | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY59174
Encompass_Preferred
5-Bromothiophene-2-carbaldehyde is a biochemical reagent that serves as a biological material or organic compound for life science research. It has applications in preparing 5-[18F]fluoro-2-2-thiophene carboxaldehyde and in biological studies assessing the anti-inflammatory and anti-tumor activity of the marine mangrove *Rhizophora apiculata*.
  • Biochemical reagent for life science research
  • Used in the preparation of 5-[18F]fluoro-2-2-thiophene carboxaldehyde
  • Applicable in anti-inflammatory studies
  • Applicable in anti-tumor activity studies
Catalog No. 50-003-58701