Learn More
Medchemexpress LLC 5-carboxamidotryptamine maleate | 74885-72-6 | 99.0% | 319.31 | C15H17N3O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10094210MG
Encompass_Preferred
5-Carboxamidotryptamine maleate is a research-grade tryptamine derivative that acts as a potent agonist at multiple serotonin (5-HT) receptor subtypes, including 5-HT1A, 5-HT1B, 5-HT1D, 5-HT5, and 5-HT7. Supplied as a solid maleate salt, it is used in pharmacology and neuroscience studies to probe 5-HT receptor function in vitro and in vivo.
  • High purity suitable for research applications.
  • Solid powder form for easy weighing and storage.
  • Stable as maleate salt for improved handling.
  • Detailed storage recommendations for long-term stability.
  • Available in small milligram quantities for screening studies.
Catalog No. 50-002-12296