Learn More
Medchemexpress LLC 5-Carboxy-2'-deoxyuridine | 14599-46-3 | MFCD22495153 | 99.0% | 272.21 g/mol | C10H12N2O7 | 5 MG

Supplier: Medchemexpress LLC HYW4247795MG
5-Carboxy-2'-deoxyuridine is a nucleoside derivative - a methyl oxidation product of thymidine and a metabolite of trifluridine. It is supplied as a research standard for analytical and biochemical studies.
- Metabolite of trifluridine and methyl oxidation product of thymidine.
- Molecular formula C10H12N2O7; molecular weight 272.21 g/mol.
- Available in small research quantities: 1 mg, 5 mg, 10 mg.
- High purity (~98.98%); contact manufacturer for certificate of analysis and SDS.
- Intended for research use only, not for human or animal therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.