Learn More
Apexbio Technology LLC 5-CFDA 79955-27-4 50mg
SDP
Supplier:  Apexbio Technology LLC A467350
Encompass_Preferred
5-CFDA (CAS 79955-27-4) is a fluorescein derivative used as a cell-permeable probe in biomedical research 5-CFDA is designed to diffuse across cellular membranes and upon entry is enzymatically hydrolyzed by intracellular esterases to generate the polar fluorescent metabolite 5-carboxyfluorescein which is retained within viable cells 5-CFDA exerts its activity primarily through intracellular esterase-mediated hydrolysis In cell-based studies 5-CFDA demonstrates efficient cell labeling and retention facilitating visualization Based on these properties 5-CFDA has research potential in cell tracking viability assays proliferation studies and live-cell imaging analyses including applications in fluorescence-based visualization of human intervertebral disc cells and related cell and tissue imaging research
Catalog No. 50-000-93438