Learn More
Medchemexpress LLC 5-chloro-2-[[2-[2-[4-(diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]-benzoic acid, monosodium salt | 1103926-82-4 | 99.0% | 543.97 | C28H27ClN3NaO5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10044710MM 1ML
Encompass_Preferred
TM5275 sodium is the sodium salt form of a small-molecule plasminogen activator inhibitor-1 (PAI-1) inhibitor supplied as a ready-to-use solution for research applications. It exhibits micromolar potency against PAI-1 and is intended for in vitro and preclinical research use only.
  • Inhibits plasminogen activator inhibitor-1 (PAI-1) with an IC50 of 6.95 μM.
  • Supplied as a 10 mM solution in DMSO (1 mL).
  • Provided as the sodium salt for improved solubility.
  • High purity (≈99.0%) suitable for research assays.
  • Molecular weight 543.97 g/mol.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-14728