Learn More
Medchemexpress LLC 5-chloro-2-(4-chlorophenyl)-4-methyl-6-(3-(piperidin-1-yl)propoxy)pyrimidine | 1639220-19-1 | 98.6% | 380.31 | C19H23Cl2N3O | 10MG
SDP
Supplier:  Medchemexpress LLC HY12582110MG
Encompass_Preferred
Sigma-1 receptor antagonist 1 is a potent, selective antagonist of the sigma-1 (σ1) receptor used in preclinical pharmacology and neuropathic pain research. The compound is supplied as a light yellow solid with high chemical purity and is characterized by the molecular formula C19H23Cl2N3O and molecular weight 380.31.
  • Potent, selective sigma-1 receptor antagonist (Ki = 1.06 nM).
  • Exhibits antineuropathic pain activity in preclinical models.
  • High purity: 98.6%.
  • Solid, light yellow to yellow appearance for easy handling.
  • Molecular formula C19H23Cl2N3O; molecular weight 380.31.
  • Supplied in small research sizes suitable for in vitro and in vivo studies.
Catalog No. 50-002-35434