Learn More
Medchemexpress LLC 5-chloro-2-mercaptobenzimidazole | 25369-78-2 | MFCD01658762 | 99.8% | 184.65 | C7H5ClN2S | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03797610G
Encompass_Preferred
Tyrosinase-IN-22 (5-Chloro-2-mercaptobenzimidazole) is a potent small-molecule inhibitor of tyrosinase substrates used in biochemical and cell-based research. It exhibits IC50 values of 60 nM (L-tyrosine) and 30 nM (L-dopa), is supplied as a white to off-white solid, and is highly soluble in DMSO. Recommended storage and formulation guidance are provided in the product documentation.
  • Potent inhibition of tyrosinase substrates at low-nanomolar concentrations.
  • High purity confirmed by certificate of analysis.
  • White to off-white solid with high DMSO solubility (≈200 mg/mL).
  • Formulation guidance provided for in vivo use (DMSO/PEG300/Tween-80 or DMSO/corn oil).
  • Stable under recommended storage: powder at -20°C, solutions at -80°C for long term.
Catalog No. 50-002-99755