Learn More
Medchemexpress LLC 5α-cholestan-3β-ol-d5 | 2260669-14-3 | 99.2% | 393.70 g/mol | C27H43D5O | 1 MG

Supplier: Medchemexpress LLC HY107819S1MG
5α-Cholestan-3β-ol-d5 is the deuterium-labeled analogue of 5α-cholestan-3β-ol, provided as a stable isotope standard for analytical chemistry. It is intended for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS workflows and is supplied with high chemical and isotopic purity.
- High isotopic and chemical purity suitable for quantitative analysis.
- Suitable as a tracer or internal standard in NMR, GC-MS, and LC-MS.
- Molecular weight 393.70 g/mol.
- Chemical formula C27H43D5O.
- Supplied in small, laboratory-scale quantities for analytical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.