Learn More
Chondrex Inc Arthrogen-CIA 5-Clone Cocktail Kit, Rapid Induction of CAIA, Broad Cross-Reactivity, 100 mg
Supplier:  Chondrex Inc 53100
Encompass
The Arthrogen-CIA 5-Clone Cocktail Kit is a specialized reagent for inducing Collagen Antibody-Induced Arthritis (CAIA) in mice, enabling rapid and consistent arthritis onset. This kit features a standardized cocktail of five monoclonal antibodies with broad cross-reactivity to various species' collagen, ensuring reproducibility and high research-grade purity. It's essential for studies in inflammatory arthritis, differentiating itself through its reliable composition and diverse application.
Catalog No. NC1713176
Add to Cart
Request a Quote