Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Cyano-2,3-di-(p-tolyl)tetrazolium chloride (CTC) | 90217-02-0 | 98.1% | 1 MG
SDP

Supplier:  Medchemexpress LLC HYD00161MG

Encompass_Preferred

5-Cyano-2,3-di-(p-tolyl)tetrazolium chloride (CTC) is a redox-sensitive red fluorescent dye that can be used for detecting metabolic activity in microorganisms. The emission maximum is 602 nm.

  • Fluorescent dye
  • Redox-sensitive
  • Emission maximum of 645 nm
  • Excitation maximum of 602 nm
  • Solid appearance
  • White to off-white color
  • Used for detecting metabolic activity in microorganisms

Catalog No. 50-004-09698


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.