Learn More
Medchemexpress LLC 5-cyclopropyl-2-[1-[(2-fluorophenyl)methyl]pyrazolo[5,4-b]pyridin-3-yl]pyrimidin-4-amine | 256376-24-6 | MFCD06411391 | 99.6% | 360.39 g/mol | C20H17FN6 | 25MG

Supplier: Medchemexpress LLC HY1237625MG
This compound is an orally active soluble guanylate cyclase (sGC) activator used in research to increase intracellular cyclic GMP (cGMP) and probe NO-independent sGC signaling. It is supplied as a white to off-white solid suitable for in vitro and in vivo studies, with recommended solubility and storage conditions for reproducible handling.
- Purity 99.6%.
- Molecular formula C20H17FN6; molecular weight 360.4 g/mol.
- Solid form (white to off-white) for powder handling and storage.
- Soluble in DMSO (17.5 mg/mL) for in vitro applications.
- In vivo formulation options include 10% DMSO/40% PEG300/5% Tween-80/45% saline and other vehicles.
- Recommended in vitro concentration 10 μM to increase cGMP in A7r5 cells.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.