Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-cyclopropyl-4-(piperidin-4-ylmethoxy)-2-fluoro-N-methylsulfonylbenzamide (GDC-0310) | 1788063-52-4 | 98.8% | C25H29Cl2FN2O4S | 10MG
SDP

Supplier:  Medchemexpress LLC HY13908110MG

Encompass_Preferred

GDC-0310 is a selective acyl-sulfonamide inhibitor of the voltage-gated sodium channel Nav1.7 (hNav1.7), reported with subnanomolar potency. It is provided as a research compound with supplier-specified purity, recommended storage, and solubility guidance for in vitro and preclinical use.

  • Selective Nav1.7 inhibitor (IC50 = 0.6 nM).
  • Acyl-sulfonamide chemotype.
  • Molecular formula C25H29Cl2FN2O4S.
  • High purity (≈98.8%).
  • White to off-white solid appearance.
  • Soluble in DMSO; may require warming or sonication.
  • Recommended storage: powder at -20°C; in solution store at -80°C for long term.
  • Intended for research use only.

Catalog No. 50-002-61690


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.