Learn More
Cayman Chemical 5-deoxy ThymIdIn 10mg
SDP
Supplier:  Cayman Chemical 1815410 MG
Encompass_Preferred
A form of thymidine in which the hydroxyl group on carbon 5 of ribose has been replaced with hydrogen and cannot be phosphorylated; competitively inhibits the cellular influx of thymidine
Catalog No. 50-297-9821