Learn More
Medchemexpress LLC 5-difluoromethoxy-2-mercaptobenzimidazole | 97963-62-7 | MFCD00467504 | 99.6% | 216.21 | C8H6F2N2OS | 25 G

Supplier: Medchemexpress LLC 25GHYW040299
5-Difluoromethoxy-2-mercaptobenzimidazole is a benzimidazole derivative supplied as a high-purity analytical reagent and synthetic intermediate for life-science research and chemical synthesis. It is provided as a white to off-white solid suitable for analytical, reference standard, and synthetic applications.
- High purity (99.6%).
- Molecular weight 216.21 g·mol-1.
- Molecular formula C8H6F2N2OS.
- Appearance: solid, white to off-white.
- Typical package: 25 g.
- Applications include use as an analytical standard and intermediate in organic synthesis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.