Learn More
Medchemexpress LLC 5'-DMT-3'-TBDMS-Bz-rA | 81256-88-4 | 99.2% | 787.97 g/mol | C44H49N5O7Si | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0181791G
Encompass_Preferred
5'-DMT-3'-TBDMS-Bz-rA is a protected ribonucleoside used in oligonucleotide synthesis to protect and modify the ribose and adenine base during chemical assembly. The combination of 5'-dimethoxytrityl (DMT), 3'-tert-butyldimethylsilyl (TBDMS), and benzoyl (Bz) protections facilitates selective reactions, handling, and downstream manipulations in synthetic workflows.
  • Provides 5' DMT, 3' TBDMS, and base benzoyl protections.
  • Designed for oligonucleotide and nucleoside chemistry applications.
  • High purity suitable for research use.
  • Solid, white to off-white appearance for easy handling.
  • Store protected from light at 4°C; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in small research pack sizes (e.g., 250 mg, 1 g).
Catalog No. 50-003-01627