Learn More
Medchemexpress LLC 5-ethenyl-9,10-dihydro-1-methyl-2,7-phenanthrenediol | 73166-28-6 | 99.8% | 252.31 g/mol | C17H16O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN513010MG
Encompass_Preferred
Effusol is a natural phenolic constituent isolated from Juncus effusus with potent radical-scavenging antioxidant activity and reported neuroprotective effects. Supplied as a research-grade solid, the manufacturer provides solubility guidance and storage recommendations for experimental use.
  • Natural phenolic constituent from Juncus effusus.
  • Potent antioxidant activity against DPPH and ABTS radicals.
  • Reported neuroprotective effect rescuing corticosterone-attenuated CA1 long-term potentiation.
  • Provided in small research sizes suitable for in vitro and in vivo studies.
  • Includes solubility guidance and recommended storage conditions for handling.
Catalog No. 50-002-88938