Learn More
Cayman Chemical 5-Ethynyl-2'-deoxyuridine, 61135-33-9, 50 mg
SDP
Supplier:  Cayman Chemical 2051850
Encompass_Preferred
Molecular Formula C11H12N2O5, Purity ≥98%, Formula Weight 252.2, Synonyms: 2'-deoxy-5-ethynyl-uridine, 5-EdU, EdU, 2'-deoxy-5-Ethynyluridine; A nucleoside analog of thymidine that is used to monitor de novo DNA synthesis through click chemistry; can be detected using a fluorescent azide probe and copper catalysis using 1,3-dipolar cycloaddition.
Catalog No. NC1275429