Learn More
Abcam 5-Ethynyluridine (5-EU), Uridine analog, 10MG
Supplier:  Abcam AB14664210MG
Encompass_Preferred
MW 268.22 Da, Purity >99%. Uridine (ab143255) analog. Click chemistry RNA transcription label. Incorporates into RNA during active RNA synthesis. Does not incorporate into DNA. Reacts with azide-containing detection reagents.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3224