Learn More
Medchemexpress LLC 5-FAM-PMDM6 | 97.02% | 1784.23 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP53671MG
Encompass_Preferred
This bioactive peptide serves as a fluorescent-labeled probe for MDM2-binding assays. It is intended for research use only and has a purity of 97.02%. This product is supplied as a light yellow to yellow solid.
  • Biological active peptide
  • Fluorescent-labeled probe for MDM2-binding assay
  • For research purposes
Catalog No. 50-003-99694