Learn More
Medchemexpress LLC 5-fluoro-1,3-dimethyl-N-[2-(4-methylpentan-2-yl)phenyl]pyrazole-4-carboxamide | 494793-67-8 | MFCD22200854 | C18H24FN3O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13916110MG
Encompass_Preferred
Penflufen is a succinate dehydrogenase inhibitor (SDHI) fungicide provided as an analytical standard for laboratory research. The compound (CAS 494793-67-8) is supplied as a solid reference material with high reported purity, intended for use in biochemical assays, analytical method development, formulation studies, and fungicide efficacy research.
  • High purity (99.8%) appropriate for analytical use.
  • Molecular formula C18H24FN3O; molecular weight 317.40 g/mol.
  • Succinate dehydrogenase inhibitor activity against diverse fungal pathogens.
  • Available in small pack sizes suited to bench-scale experiments.
  • Includes batch certificate of analysis and HPLC data for verification.
  • Suitable as a reference standard for assay development and formulation testing.
Catalog No. 50-002-61069