Learn More
Medchemexpress LLC 5-fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione | 2230957-36-3 | >98.0% | 290.25 | C14H11FN2O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW577257500MG
Encompass_Preferred
5-Fluoro-2-(1-methyl-2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione is a fluorinated thalidomide analogue described as an E3 ligase activator for research use. Supplied as a powder, it can serve as a synthetic building block for further derivatization, particularly by nucleophilic aromatic substitution (SNAr).
  • Fluorinated thalidomide analogue useful for E3 ligase activation studies.
  • Suitable as a synthetic building block for derivatization via SNAr.
  • Supplied as a powder suitable for laboratory handling and storage.
  • High purity; commonly listed as >98.0%.
  • Typical pack sizes: 100 mg-1 g.
Catalog No. 50-002-84428