Learn More
Medchemexpress LLC 5-Fluoro-2-(1H-pyrrol-1-yl)aniline | 896429-57-5 | 98.3% | 176.19 | 500 MG

Supplier: Medchemexpress LLC HY23660500MG
5-Fluoro-2-(1H-pyrrol-1-yl)aniline is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds.
- Used in the synthesis of active compounds.
- Appearance is brown to dark brown solid.
- 1H NMR spectrum consistent with structure.
- LCMS consistent with structure.
- Purity of 98.3% (LCMS).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.