Learn More
Medchemexpress LLC 5-fluoro-2-methyl-N-[4-(5H-pyrrolo[2,1-c]-benzodiazepin-10-ylcarbonyl)-3-chlorophenyl]benzamide | 168079-32-1 | MFCD00937905 | 99.7% | 473.93 | C27H21ClFN3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY141855MG
Encompass_Preferred
Lixivaptan is an orally active, selective vasopressin V2 receptor antagonist supplied for research use. It is commonly used as a pharmacological tool to study V2 receptor signaling and ligand-receptor interactions. The compound is provided as a solid in multiple pack sizes and as a 10 mM solution in DMSO, and is characterized by high purity and documented activity data.
  • Orally active selective vasopressin V2 receptor antagonist.
  • High potency (human V2 IC50 ≈ 1.2 nM).
  • High purity (approximately 99.7%).
  • Supplied as solid and as 10 mM solution in DMSO.
  • Available in multiple small pack sizes for research use.
  • Storage: powder at 4°C; in solvent -80°C for long term.
  • Useful for receptor pharmacology and signaling studies.
Catalog No. 50-002-21354