Learn More
Medchemexpress LLC 5-fluoro-2-[(Z)-(2-hexahydropyridazin-1-yl-4-oxo-thiazol-5-ylidene)methyl]phenyl pyrrolidine-1-carboxylate | 1181083-81-7 | ≥98.0% | 404.46 g/mol | C19H21FN4O3S | 1 ML

Supplier: Medchemexpress LLC HY10302310MM 1ML
CLP290 is a small-molecule activator of the neuron-specific K+-Cl- cotransporter KCC2 used in neuroscience research. It is provided as a 10 mM solution in DMSO for laboratory use.
- Small-molecule activator of the neuron-specific K+-Cl- cotransporter KCC2.
- Used for neuroscience research exploring neuronal inhibition and pain models.
- Supplied as a 10 mM solution in DMSO, 1 mL.
- Molecular formula C19H21FN4O3S; molecular weight 404.46 g/mol.
- Purity ≥98.0% (HPLC).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.