Learn More
Cayman Chemical 5-fluoro nI 1mg
SDP
Supplier:  Cayman Chemical 141471 MG
Encompass_Preferred
A derivative of NNEI in which a fluorine atom has been added to the terminal carbon of the pentyl chain; intended for forensic and research applications
Catalog No. 50-295-4407