Learn More
Selleck Chemical LLC 5-Fluoroorotic acid S9873-100MG
SDP
Supplier:  Selleck Chemical LLC S9873100MG
Encompass_Preferred
5-Fluoroorotic acid (5-FOA 5-Fluoroorotate NSC 31712) is a selective agent in yeast molecular genetics 5-Fluoroorotic acid possesses a well-expressed anticandidal effect close to that of 5-fluorocytosine as well as moderate antidermatophytal effects
Catalog No. 50-000-23800