Learn More
Medchemexpress LLC 5-fluoroorotic acid | 703-95-7 | MFCD00042526 | 99.4% | 174.09 g/mol | C5H3FN2O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW016819500MG
Encompass_Preferred
5-Fluoroorotic acid is a pyrimidine analog that inhibits thymidylate synthase and is used as a selective agent in yeast molecular genetics. It is provided for research use and has reported antimalarial activity.
  • Thymidylate synthase inhibitor used in biochemical research.
  • Selective agent for yeast genetics applications.
  • Reported antimalarial activity documented in literature.
  • Available in solid and solution formats for experimental flexibility.
  • High purity suitable for analytical and biological assays.
Catalog No. 50-002-99962