Learn More
Medchemexpress LLC 5-fluorouridine 5'-phosphate | 796-66-7 | MFCD30532520 | 98.6% | 342.17 g·mol⁻¹ | C9H12FN2O9P | 100mg
SDP
Supplier:  Medchemexpress LLC HYW784565100MG
Encompass_Preferred
5-Fluorouridine 5 -phosphate is an ODCase (orotidine-5 -monophosphate decarboxylase) inhibitor with a Ki value of 98 M for human ODCase and a Ki value of 645 M for Methanobacterium ODCase 5-Fluorouridine 5 -phosphate exhibits inhibitory activity against leukemia and lymphoma cell lines and can be used in cancer research [1]
Catalog No. 50-002-69016