Learn More
Alkali Scientific 5 FOA [5 Fluoroorotic acid], 5 G
SDP
Supplier:  Alkali Scientific FOA005CP
Encompass
Fluoroorotic acid (FOA) is a key reagent used in nucleic acid selection, particularly for the genetic modification of yeast and bacteria. The 5g package is ideal for smaller-scale experiments and laboratory applications requiring the selective elimination of uracil-containing cells. By converting 5-fluorouracil (5-FU) into a toxic nucleotide, FOA effectively kills non-transformed cells, making it an essential tool for molecular biology research, particularly in genetic research and drug discovery. The 5g quantity is suitable for researchers needing a cost-effective amount for selection processes, ensuring efficient identification and isolation of genetically modified or recombinant cells.
Catalog No. 50-006-61865