Learn More
Cayman Chemical 5-FormyluracIl 500mg
SDP
Supplier:  Cayman Chemical 36523500 MG
Encompass_Preferred
An oxidized form of thymine and a marker of DNA oxidative damage; incorporation of 5-formyluracil into DNA forms mispairings with thymine, cytosine, and guanine and induces mutations in E. coli; decreases the viability of V9-4 fibroblasts at 1 mM and increases their mutation frequency in a concentration-dependent manner
Catalog No. 50-291-2049