Learn More
AdipoGen 5-Geranyloxy-7-methoxycoumarin
SDP
Supplier:  AdipoGen CDXG0228M001
Encompass_Preferred
Chemical. CAS 7380-39-4. Formula C20H24O4. MW 328.4. 5-Geranyloxy-7-methoxycoumarin is a natural coumarin found in the essential oils of citrus such as bergamot, which shows antifungal, antibacterial, and anticancer activity. 5-Geranyloxy-7-methoxycoumarin arrests cells at the G0/G1 phase and induces apoptosis through activation of tumour suppressor gene p53, caspase8/3, regulation of Bcl2 and inhibition of p38 MAPK phosphorylation.
Catalog No. 50-256-0814