Learn More
AdipoGen 5-(Hydroxymethyl)-2'-deoxycytidine (5 mg)
SDP
Supplier:  AdipoGen AGCR13532M005
Encompass_Preferred
Chemical. CAS: 7226-77-9. Formula: C10H15N3O5. MW: 257.2. Synthetic. Epigenetic base. A modified pyrimidine that is capable of producing interstrand cross-links in double-stranded DNA and has been used to quantify DNA hydroxymethylation levels in biological samples. Enriched in the brain, suggesting a role in epigenetic control of neuronal function. Used in epigenetic research and important for cancer research. Potent HIV-1 reverse transcriptase activity inhibitor. Can be used as building block in nucleoside and nucleotide chemistry.
Catalog No. 50-205-3923