Learn More
AdipoGen 5-Hydroxy-2-methyl-4-chromanon
SDP
Supplier:  AdipoGen BVT0322M001
Encompass_Preferred
Chemical. CAS 14153-17-4. Formula C10H10O3. MW 178.2. Isolated from Nodulisporium sp. Antifungal and phytotoxic. Inhibits the growth of Saccharomyces cerevisiae and the formation of soybean callus.
Catalog No. 50-256-1990